PYREX® Volumetric Flasks, Class A, Glass, Clear, NS Glass Stopper, Corning
Supplier: Corning
Certificates
About this item
PYREX® borosilicate glass.
Flasks feature sharp, permanent graduation line and large white block letter inscriptions. Body is machine-blown with sealed heavy-tubing neck for increased strength. Hollow glass ground NS stoppers, where included, are interchangeable.
All flasks are graduated to Class A volumetric tolerances and calibrated to meet ASTM specifications E-694, E-542, and E-288. Certified flasks are individually serialized and supplied with NIST-traceable certificate of identification and capacity.
Specifications
- Catalog No:
- 29624-015
- 29624-102
- 29624-123
- 29624-167
- 29624-065
- 29624-043
- 29624-087
- 29624-145
- 29624-022
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
Specifications
Related Information
Certificates
Search by Lot Number29624-015_19520016 29624-022_08119048 29624-043_13520023 29624-043_28119005 29624-123_33719014
Conformity declaration
More
Customers Who Bought This Also Bought