About this item
High vacuum grease is ideal for sealing and lubricating vacuum and pressure systems
This silicone product has a low volatility to hold deep vacuums; it resists oxidation, is nonmelting and nongumming, and has good thermal stability. Grease is translucent white with a heavy consistency.
It can also be used to maintain laboratory equipment.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- CA59344-066
- CA59344-055
- Temperature range:
- –40 to 204°C (–40 to 400°F)
- –40 to 204°C (–40 to 400°F)