2-Propanol ≥99.5% (by GC), GR ACS, Supelco®
Supplier: MilliporeSigma
Certificates
About this item
Synonyms: IPA,Isopropyl Rubbing Alcohol,Isopropyl alcohol,Dimethyl carbinol,2-Hydroxy propane,Isopropanol
Formula:(CH₃)₂CHOH
MW:60.1 g/mol
Boiling Pt:82 °C (1013 hPa)
Melting Pt:–89 °C
Density:0.786 g/cm³ (20 °C)
Flash Pt:12 °C
MDL Number:MFCD00011674
CAS Number:67-63-0
ADR:3,II
Merck Index:13,05228
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,5
:Boiling Pt:
:Boiling Pt: 82 °C (1013 hPa)
:Melting Pt:
:Melting Pt: -89 °C|Melting Pt: –89 °C
:Flash Pt:
:Flash Pt: 12 °C
:M.W. 60.1 g/mol
:2-Propanol
EINECS:200-661-7
UN:1219
REACH:01-2119457558-25
:3
:II
:GHS02
:GHS07
Storage Temperature:Ambient
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Assay (GC):99.5% min
- Carbonyl compounds (acetone or propionaldehyde):0.002% max
- Color (APHA):10 max
- Form:Clear liquid
- Infrared spectrum:Conforms to standard
- Residue after evaporation:0.001% max
- Solubility in water:To pass test
- Titrable acid:0.0001meq/g max
- Titrable base:0.0001meq/g max
- Water (H2O):0.2% max
- Supelco®, 2-Propanol ≥99.5% (by GC), GR ACS:
Specification Test Results
Related Information
Certificates
Search by Lot NumberEMD Millipore Start and End with Quality Product Portfolio Brochure EMD Millipore Sustainable Products Brochure VWR® Chemicals in Poly-Coated Glass Bottles
Product Literature
VWR GHS Compliance Letter (US)
Additional Product Details
Customers Who Bought This Also Bought