About this item
NEXTFLEX™ human whole exome sequencing (WES) panel is a comprehensive solution that covers all target regions of major WES panels available in the market.
- Complete whole exome coverage
- Exceptional target capture performance
- Rapid same-day workflow
- Automated on Sciclone® G3 NGSx workstation
- Bead-based normalization
The NEXTFLEX™ WES panel provides exceptional target capture performance due to probe design and reagent optimization technology. Despite some companies who resort to masking the hard-to-capture regions (such as GC- or AT-rich regions and homologous regions) or completely omit the regions from their target in order to enhance the quality of their results, the NEXTFLEX™ WES panel provides both high coverage and on-target ratio without reducing the number of target regions. NEXTFLEX™ WES panel captures regions that other panels cannot capture with quality coverage and uniformity. The all-around performance of the NEXTFLEX™ WES panel allows for cost-effective and time-saving sequencing of the whole exome.