Nitric acid 68.0 - 70.0%, AR Select® ACS for trace metal analysis, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Non-Regulated, With Information
About this item
Formula:HNO₃
MW:63.01 g/mol
Boiling Pt:120.5 °C (1013 hPa)
Melting Pt:–42 °C
Density:1.4 g/cm³ (20 °C)
MDL Number:MFCD00011349
CAS Number:7697-37-2
Merck Index:13,06608
:63.01
ADR:8,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Management of Change (MOC) category = NI
:Boiling Pt:
:Boiling Pt: 120.5 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –42 °C
:Flash Pt:
:Flash Pt:
:Density: 1.51 g/cm³ (20 °C)
:M.W. 63.01 g/mol
:Nitric acid
Storage Temperature:Ambient
EINECS:231-714-2
UN:2031
:8
:II
:1.51
:GHS03
:GHS05
:GHS07
:M.O.C. Category NI
:12,06672
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Meets ACS Reagent Chemical Requirements:
- For Laboratory,Research,or Manufacturing Use:
- ACS - Assay (HNO₃):68.0 - 70.0 %
- Appearance:Passes Test
- ACS - Color (APHA):≤ 10
- ACS - Arsenic (As):≤ 0.010 ppm
- ACS - Arsenic and Antimony (as As):≤ 0.005 ppm
- ACS - Chloride (Cl):≤ 0.04 ppm
- ACS - Heavy Metals (as Pb):≤ 0.05 ppm
- ACS - Residue after Ignition:≤ 5 ppm
- ACS - Sulfate (SO₄):≤ 0.4 ppm
- Determine by Flame Photometry & ICP - Aluminum (Al):≤ 50.0 ppb
- Determine by Flame Photometry & ICP - Barium (Ba):≤ 1.0 ppb
- Determine by Flame Photometry & ICP - Beryllium (Be):≤ 1.0 ppb
- Determine by Flame Photometry & ICP - Bismuth (Bi):≤ 10 ppb
- Determine by Flame Photometry & ICP - Boron (B):≤ 10.0 ppb
- Determine by Flame Photometry & ICP - Cadmium (Cd):≤ 1.0 ppb
- Determine by Flame Photometry & ICP - * Calcium (Ca):≤ 200.0 ppb
- Determine by Flame Photometry & ICP - Chromium (Cr):≤ 10.0 ppb
- Determine by Flame Photometry & ICP - Cobalt (Co):≤ 5.0 ppb
- Determine by Flame Photometry & ICP - Copper (Cu):≤ 1.0 ppb
- Determine by Flame Photometry & ICP - Gallium (Ga):≤ 20.0 ppb
- Determine by Flame Photometry & ICP - Germanium (Ge):≤ 10.0 ppb
- Determine by Flame Photometry & ICP - Gold (Au):≤ 10.0 ppb
- Determine by Flame Photometry & ICP - Iron (Fe):≤ 40.0 ppb
- Determine by Flame Photometry & ICP - Lead (Pb):≤ 1.0 ppb
- Determine by Flame Photometry & ICP - Lithium (Li):≤ 5.0 ppb
- Determine by Flame Photometry & ICP - Magnesium (Mg):≤ 10.0 ppb
- Determine by Flame Photometry & ICP - Manganese (Mn):≤ 1.0 ppb
- Determine by Flame Photometry & ICP - Mercury (Hg):≤ 1.0 ppb
- Determine by Flame Photometry & ICP - Molybdenum (Mo):≤ 5.0 ppb
- Determine by Flame Photometry & ICP - Nickel (Ni):≤ 5.0 ppb
- Determine by Flame Photometry & ICP - Potassium (K):≤ 20.0 ppb
- Determine by Flame Photometry & ICP - * Silicon (Si):≤ 50.0 ppb
- Determine by Flame Photometry & ICP - Silver (Ag):≤ 5.0 ppb
- Determine by Flame Photometry & ICP - * Sodium (Na):≤ 500.0 ppb
- Determine by Flame Photometry & ICP - Strontium (Sr):≤ 1.0 ppb
- Determine by Flame Photometry & ICP - Thallium (Tl):≤ 20.0 ppb
- Determine by Flame Photometry & ICP - Tin (Sn):≤ 10.0 ppb
- Determine by Flame Photometry & ICP - Vanadium (V):≤ 1.0 ppb
- Determine by Flame Photometry & ICP - Zinc (Zn):≤ 5.0 ppb
- Determine by Flame Photometry & ICP - Zirconium (Zr):≤ 10 ppb
- Determine by Flame Photometry & ICP:
- * May change over time due to extraction from glass container.:
- Macron Fine Chemicals™, Nitric acid 68.0 - 70.0%, AR Select® ACS for trace metal analysis:
Specification Test Results
Related Information
Certificates
Search by Lot Number6623 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
sds_MACR6623
SDS
spec_MK662346
Product specification
Customers Who Bought This Also Bought