Nitric acid 68.0 - 70.0% (by acidimetry), AR® ACS, Macron Fine Chemicals™
Supplier: Avantor
MOC: Non-Regulated, With Information
About this item
Formula:HNO₃
MW:63.01 g/mol
Boiling Pt:120.5 °C (1013 hPa)
Melting Pt:–42 °C
Density:1.4 g/cm³ (20 °C)
MDL Number:MFCD00011349
CAS Number:7697-37-2
Merck Index:13,06608
:63.01
ADR:8,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Management of Change (MOC) category = NI
:Boiling Pt:
:Boiling Pt: 120.5 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –42 °C
:Flash Pt:
:Flash Pt:
:Density: 1.51 g/cm³ (20 °C)
:M.W. 63.01 g/mol
:Nitric acid
Storage Temperature:Ambient
EINECS:231-714-2
UN:2031
:8
:II
:1.51
:GHS03
:GHS05
:GHS07
:M.O.C. Category NI
:12,06672
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Meets ACS Reagent Chemical Requirements:
- For Laboratory,Research,or Manufacturing Use:
- Appearance:Passes Test
- ACS - Assay (HNO₃) (by acidimetry):68.0 - 70.0 %
- ACS - Color (APHA):≤10
- ACS - Residue after Ignition:≤5 ppm
- ACS - Chloride (Cl):≤0.5 ppm
- ACS - Sulfate (SO₄):≤1 ppm
- ACS - Arsenic (As):≤0.010 ppm
- ACS - Heavy Metals (as Pb):≤0.2 ppm
- Trace Impurities - ACS - Iron (Fe):≤0.200 ppm
- Macron Fine Chemicals™, Nitric acid 68.0 - 70.0% (by acidimetry), AR® ACS:
Specification Test Results
Related Information
Certificates
Search by Lot Number1409 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Macron 1409.pdf
Datasheet
sds_MACR1409
SDS
Customers Who Bought This Also Bought