DOPlify™ WGA V2 Kits
Catalog #: CA77448-068
Supplier: REVVITY HEALTH SCIENCES CANADA CA
New Product
CAS Number:
Specifications
- Description:DOPlify™ WGA v2 kit
- No. of Reactions:50
Specifications
About this item
The DOPlify® WGA V2 kit performs fast whole genome amplification on single cells or limited template DNA samples, allowing for the cell chromosome copy number status to be determined.
- NGS compatible advanced DOP-PCR
- Fast whole genome amplification with protocol time <3 hours
- Reduced contamination risk with only two tube openings required and three reagents to add
- Can be fully automated with the JANUS® G3 liquid handler
- No amplification of no template control
- 2.5 μg of DNA from one cell
The DOPlify® WGA V2 kit offers the advantage of full mitochondrial DNA coverage and the ability to add custom primers to specially amplify regions of interest. Products of the DOPlify® WGA V2 kit can be used for a wide range of downstream processes, workflows, and NGS applications. The technology captures exclusive know-how for the lysis and whole genome amplification, with reagents provided in an easy-to-use kit format and less than three hours protocol time.
Certifications: Manufactured in ISO 9001 facility.
Caution: For research use only. Not for use in diagnostic procedures.
Customers Who Bought This Also Bought