Dial Thermometers, 1.75/2" Diameter
Supplier: Heathrow Scientific
Certificates
About this item
Our dial thermometers are an excellent choice for many applications including food and beverage quality control, general lab use, and asphalt/concrete testing.
- Stem length 203 mm (8")
Each of these dial thermometers feature an 8" stem and come with a clip for attaching to containers, tanks, etc.
Customers Who Bought This Also Bought