Triethanolamine (trolamine) 99.0-103.0% (by acidimetry), SAFC®
Supplier: MilliporeSigma
Certificates
About this item
Synonyms: Tris-(2-hydroxyethyl)amine,2,2'
Formula:C₆H₁₅NO₃
MW:149.19 g/mol
Boiling Pt:335.4 °C (1013 hPa)
Melting Pt:21.6 °C
Density:1.13 g/cm³ (20 °C)
Flash Pt:179 °C (closed cup)
MDL Number:MFCD00002855
CAS Number:102-71-6
Merck Index:12,09798
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,0-103,0
:Boiling Pt:
:Boiling Pt: 335.4 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 21.6 °C
:Flash Pt:
:Flash Pt: 179 °C (closed cup)
:M.W. 149.19 g/mol
EINECS:203-049-8
:179 °C
Storage Temperature:Ambient
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Assay (acidimetric, calc. on anhydrous substance):99.0 - 103.0 %
- Identity (IR-spectrum):passes test
- Identity (GC):conforms
- Appearance:colorless to slightly yellow viscose liquid
- Appearance of solution:Clear and not more intense in colour than reference solution B₆
- Clarity of solution:passes test
- PH (1.0 g in 10 ml water):10.5 - 11.5
- Refractive index (n 20/D):1.482 - 1.485
- Relative Density (d 20/20):1.120 - 1.130
- Relative Density (d 25/25):1.120 - 1.128
- Heavy metals (as Pb):≤ 0.0010 %
- As (Arsenic):≤ 0.000075 %
- Fe (Iron):≤ 0.0005 %
- Related substances (Ethanolamine, GC):≤ 0.1 %
- Related substances (Diethanolamine, GC):≤ 0.5 %
- Related substances (total impurity, GC):≤ 1.0 %
- Related substances (N-Nitrosodiethanolamine, GC):≤ 24 ppb
- Residual solvents (ICH Q3C):...excluded by production process
- Sulfated ash (600 °C):≤ 0.05 %
- Water (according to Karl Fischer):≤ 0.5 %
- Endotoxins:≤ 2.5 I.U./ml
- Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).:
- N-Nitrosodiethanolamine (GC) test performed by a qualified external laboratory.:
- Meets analytical specifications of Ph Eur, JPE, NF.:
- The information provided does not imply the suitability of the product for pharmaceutical applications. It is customer's sole responsibility, prior to use, to determine that the product is suitable and permitted for the customer's intended use and application.:
Specification Test Results
Related Information
Customers Who Bought This Also Bought