2,2,4-Trimethylpentane ≥99.5% (by GC), OmniSolv®, Supelco®
Supplier: MilliporeSigma
Certificates
About this item
Synonyms: Isobutyltrimethylmethane,Isooctane
Formula:(CH₃)₂CHCH₂C(CH₃)₃
MW:114.23 g/mol
Boiling Pt:99.2 °C (1013 hPa)
Melting Pt:–107 °C
Density:0.692 g/cm³ (20 °C)
Flash Pt:–12 °C
MDL Number:MFCD00008943
CAS Number:540-84-1
ADR:3,II
Merck Index:13,05212
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,5
:Boiling Pt:
:Boiling Pt: 99.2 °C (1013 hPa)
:Melting Pt:
:Melting Pt: -107 °C|Melting Pt: –107 °C
:Flash Pt:
:Flash Pt: –12 °C
:M.W. 114.23 g/mol
Storage Temperature:Ambient
EINECS:208-759-1
UN:1262
:3
:II
:–12 °C
:GHS02
:GHS08
:GHS07
:GHS09
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Assay (GC):99.5% min
- Capillary ECD responsive substances (as PCNB):5ng/L max
- Color (APHA):10 max
- Filtered through 0.2 µm filter:To pass test
- Fluorescence (as quinine base):250ppt max
- Form:Clear liquid
- Infrared spectrum:Conforms to standard
- Residue after evaporation:1ppm max
- UV Abs. at 200 nm:1.00AU max
- UV Abs. at 220 nm:0.05AU max
- UV Abs. at 230 nm:0.02AU max
- UV Abs. at 250 nm:0.005AU max
- UV Abs. at 270 nm:0.005AU max
- UV Abs. at 300 nm:0.005AU max
- UV Cut-off:200nm max
- Water (H2O):0.01% max
Specification Test Results
Related Information
Certificates
Search by Lot NumberVWR GHS Compliance Letter (US)
Additional Product Details
VWR® Chemicals in Poly-Coated Glass Bottles
Product Literature
Customers Who Bought This Also Bought