Trifluoroacetic acid 0.1% (v/v) in acetonitrile mobile phase for HPLC, Supelco®
Catalog #: CA-TX1277-NP20
Supplier: MilliporeSigma
CAS Number: 76-05-1
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Plastic barrel
- Size:20 L
- Cat. no.:CA-TX1277-NP20
- Supplier No.:TX1277-NP20
Specifications
Specification Test Results
- Assay (trifluoroacetic acid):0.095-0.105% (v/v)
- Color (APHA):10 max
- Filtered through 0.2 µm filter:To pass test
- Form:Clear liquid
- Gradient at 210 nm:0.005AU max
- Gradient at 220 nm:0.001AU max
- Gradient at 280 nm:0.001AU max
- Residue after evaporation:5ppm max
- UV Abs. at 190 nm:1.00AU max
- UV Abs. at 200 nm:0.50AU max
- UV Abs. at 210 nm:0.40AU max
- UV Abs. at 230 nm:0.30AU max
- UV Abs. at 250 nm:0.05AU max
- UV Abs. at 280 nm:0.010AU max
- UV Abs. at 300 nm:0.010AU max
- UV Abs. at 350 nm:0.010AU max
- in acetonitrile:
Specification Test Results
About this item
Synonyms: TFA
ADR:3,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: -15 °C
:Flash Pt:
:Flash Pt:
:M.W. 114.02 g/mol
MW:114.02 g/mol
EINECS:200-929-3
:-15
:71.8
UN:1648
:3
:II
:GHS02
:GHS07
Formula:C₂HF₃O₂
Storage Temperature:Ambient
:12,09812
MDL Number:MFCD00004169
CAS Number:76-05-1
Customers Who Bought This Also Bought