N-Methyl-2-pyrrolidone (NMP) ≥99.0%, ChromAR® for liquid chromatography, for UV spectrophotometry, Macron Fine Chemicals™
Catalog #: CAMK639208
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 872-50-4
MOC: Non-RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Glass bottle, amber, narrow mouth
- Size:4 L
- Cat. no.:CAMK639208
- Supplier No.:6392-08
Specifications
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:
- Assay (C₅H₉NO):≥99.0 %
- Chloride (Cl):≤1 ppm
- Color (APHA):≤50
- Free Amines (as CH₃NH₂):≤0.010 %
- Residue after Ignition:≤5 ppm
- Water (H₂O)(by Karl Fischer titrn):≤0.05 %
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 400 nm:≤0.010
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 350 nm:≤0.03
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 300 nm:≤0.20
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 285 nm:≤0.40
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 275 nm:≤1.00
- Appearance (clear, colorless liquid):
Specification Test Results
About this item
Synonyms: 1-Methyl-2-pyrrolidinone,1-Methyl-2-pyrrolidone
Boiling Pt:202 °C (1013 hPa)
Melting Pt:–24 °C
Density:1.033 g/cm³ (20 °C)
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,0
:Management of Change (MOC) category = N
Flash Pt:91 °C
:Boiling Pt:
:Boiling Pt: 202 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –24 °C
:Flash Pt:
:Flash Pt: 91 °C
:M.W. 99.13 g/mol
Merck Index:12,06197
Formula:C₅H₉NO
MW:99.13 g/mol
MDL Number:MFCD00003193
EINECS:212-828-1
:GHS08
:GHS07
Storage Temperature:Ambient
:M.O.C. Category N
CAS Number:872-50-4
Customers Who Bought This Also Bought