VWR® Combustion Boats, Porcelain
Supplier: VWR International
Certificates
About this item
Chemical-porcelain combustion boats feature a perforated handle and glazing inside and out, except for bottom surface.
- Chemical porcelain construction
- For use in high-temperature applications
- Features pouring lip and flat handle
- Glazed interior and exterior
For use in temperatures up to 1150°C (2102°F).
Customers Who Bought This Also Bought