PYREX® VISTA™ Flasks, Flat Bottom, Glass, Clear, with Standard Tapered Joints, Corning
Supplier: Corning
Certificates
About this item
Made of PYREX® borosilicate glass.
- These flasks have full length outer 24/40 standard tapered joint
- Wall thickness is controlled to provide balance between thermal expansion and mechanical strength for shock resistance
Reusable short neck boiling flask with flat bottom.
Certifications: ASTM E-1403
Specifications
- Catalog No:
- 89090-122
- 89090-350
- 89090-326
- 89090-388
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey