Synthware Microscale Round-Bottom Flasks, Threaded, Kemtech America
Supplier: Kemtech America
Certificates
About this item
Supplied with one compression cap, one Viton® O-ring, and one fluoropolymer resin-faced silicone rubber septum.
- Threaded neck
- Ground glass joint
- Heavy wall borosilicate construction
- Various capacities and joint sizes available
- Cap included
Threaded ground glass joint allows for the use of a threaded cap or a ground glass stopper.
Ordering information: For replacement caps, contact your Avantor representative.
Customers Who Bought This Also Bought