Methanol, anhydrous ≥99.8% (by GC, corrected for water content), BAKER ANALYZED® ACS for Karl Fischer titration, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Non-Regulated
About this item
Synonyms: Hydroxy methane,Carbinol,Wood alcohol
Formula:CH₃OH
MW:32.04 g/mol
Boiling Pt:64.6 °C (1013 hPa)
Melting Pt:–98 °C
Density:0.7918 g/cm³ (20 °C)
Flash Pt:11 °C (closed cup)
MDL Number:MFCD00004595
CAS Number:67-56-1
ADR:3,II
Merck Index:13,05984
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,8
:Management of Change (MOC) category = N
:Boiling Pt:
:Boiling Pt: 64.6 °C (1013 hPa)
:Melting Pt:
:Melting Pt: -98 °C|Melting Pt: –98 °C
:Flash Pt:
:Flash Pt: 11 °C (closed cup)
:M.W. 32.04 g/mol
Storage Temperature:Ambient
EINECS:200-659-6
UN:1230
REACH:01-2119433307-44
:3
:II
:11 °C
:GHS02
:GHS06
:GHS08
:M.O.C. Category N
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Meets ACS Reagent Chemical Requirements:
- For Laboratory,Research,or Manufacturing Use:
- ACS - Assay (CH₃OH) (by GC, corrected for water):≥99.8 %
- ACS - Substances Darkened by H₂SO₄:Passes Test
- ACS - Substances Reducing Permanganate:Passes Test
- ACS - Solubility in H₂O:Passes Test
- ACS - Color (APHA):≤10
- ACS - Water (H₂O):≤0.007 %
- ACS - Residue after Evaporation:≤0.001 %
- Carbonyl Compounds - Acetaldehyde:≤0.001 %
- Carbonyl Compounds - Acetone:≤0.001 %
- Carbonyl Compounds - Formaldehyde:≤0.001 %
- Titrable Acid (µeq/g):≤0.3
- Titrable Base (µeq/g):≤0.2
- J.T.Baker®, Methanol, anhydrous ≥99.8% (by GC, corrected for water content), BAKER ANALYZED® ACS for Karl Fischer titration:
Specification Test Results
Related Information
Certificates
Search by Lot Number9049 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Avantor Portfolio of Chemicals VWR Top Rated Guide VWR® Chemicals in Poly-Coated Glass Bottles
Product Literature
Brand Definition Baker Analyzed
Datasheet
Customers Who Bought This Also Bought