tert-Butyl methyl ether ≥99.0% (by GC, corrected for water content), BAKER ANALYZED® HPLC for HPLC, UHPLC, for spectrophotometry, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Non-Regulated
About this item
Synonyms: MTBE
Formula:(CH₃)₃COCH₃
MW:88.15 g/mol
Boiling Pt:55.2 °C (1013 hPa)
Melting Pt:–108.6 °C
Density:0.7405 g/cm³ (20 °C)
Flash Pt:–28 °C
MDL Number:MFCD00008812
CAS Number:1634-04-4
ADR:3,II
Merck Index:12,06111
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,0
:Management of Change (MOC) category = N
:Boiling Pt:
:Boiling Pt: 55.2 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –108.6 °C
:Flash Pt:
:Flash Pt: –28 °C
:M.W. 88.15 g/mol
:tert-Butyl methyl ether
Storage Temperature:Ambient
EINECS:216-653-1
UN:2398
:3
:II
:–28 °C
:GHS02
:GHS07
:GHS08
:M.O.C. Category N
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:
- Assay (C₅H₁₂O) (by GC, corrected for water):≥99.0 %
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 350 nm:≤0.01
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 280 nm:≤0.02
- Ultraviolet Absorbance (1.00-cm cell vs. water) - 254 nm:≤0.1
- Ultraviolet Absorbance (1.00-cm cell vs. water) - UV Cut-off, nm:≤215
- Fluorescence Trace Impurities, measured as Quinine Base - at 450 nm Emission:≤0.3 ppb
- Fluorescence Trace Impurities, measured as Quinine Base - at Emission Maximum for Impurities:≤1.0 ppb
- Peroxide (as H₂O₂):≤1 ppm
- Residue after Evaporation:≤3.0 ppm
- Water (by KF, coulometric):≤0.05 %
Specification Test Results
Related Information
Certificates
Search by Lot Number9042 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Avantor High-Purity Solvents and Reagents Avantor J.T.Baker® LCMS and HPLC Solvents and Reagents Avantor Portfolio of Chemicals VWR Top Rated Guide
Product Literature
JTBaker_9042_Datasheet.pdf
Datasheet
spec_JT9042-3
Product specification
Customers Who Bought This Also Bought