N-Methyl-2-pyrrolidone (NMP) ≥99.5%, ALEG® for microelectronic, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Regulated
About this item
Synonyms: 1-Methyl-2-pyrrolidinone,1-Methyl-2-pyrrolidone
Formula:C₅H₉NO
MW:99.13 g/mol
Boiling Pt:202 °C (1013 hPa)
Melting Pt:–24 °C
Density:1.033 g/cm³ (20 °C)
Flash Pt:91 °C
MDL Number:MFCD00003193
CAS Number:872-50-4
Merck Index:12,06197
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,5
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt: 202 °C (1013 hPa)
:Melting Pt:
:Melting Pt: -24 °C|Melting Pt: –24 °C
:Flash Pt:
:Flash Pt: 91 °C
:M.W. 99.13 g/mol
Storage Temperature:Ambient
EINECS:212-828-1
:GHS08
:GHS07
:M.O.C. Category R
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- For Microelectronic Use:
- Assay (C₅H₉NO):≥99.5 %
- Color (APHA):≤20
- Water (H₂O)(by Karl Fischer titrn):≤0.05 %
- Free Amines (as CH₃NH₂):≤50 ppm
- Chloride (Cl):≤0.5 ppm
- Phosphate (PO₄):≤1 ppm
- Trace Impurities - Aluminum (Al):≤100.0 ppb
- Arsenic and Antimony (as As):≤100.0 ppb
- Trace Impurities - Barium (Ba):≤100.0 ppb
- Trace Impurities - Boron (B):≤100.0 ppb
- Trace Impurities - Calcium (Ca):≤300 ppb
- Trace Impurities - Chromium (Cr):≤100.0 ppb
- Trace Impurities - Cobalt (Co):≤100.0 ppb
- Trace Impurities - Copper (Cu):≤100.0 ppb
- Trace Impurities - Gold (Au):≤100.0 ppb
- Trace Impurities - Iron (Fe):≤100.0 ppb
- Trace Impurities - Lead (Pb):≤100 ppb
- Trace Impurities - Lithium (Li):≤100.0 ppb
- Trace Impurities - Magnesium (Mg):≤100.0 ppb
- Trace Impurities - Manganese (Mn):≤100.0 ppb
- Trace Impurities - Nickel (Ni):≤100.0 ppb
- Trace Impurities - Potassium (K):≤100 ppb
- Trace Impurities - Silver (Ag):≤100.0 ppb
- Trace Impurities - Sodium (Na):≤300 ppb
- Trace Impurities - Strontium (Sr):≤100.0 ppb
- Trace Impurities - Tin (Sn):≤100 ppb
- Trace Impurities - Titanium (Ti):≤100.0 ppb
- Trace Impurities - Vanadium (V):≤100.0 ppb
- Trace Impurities - Zinc (Zn):≤100.0 ppb
- Particle Count - 1.0 µm and greater (Rion KS42AF):≤10 par/ml
Specification Test Results
Related Information
Certificates
Search by Lot Number6397 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals VWR Top Rated Guide
Product Literature
sds_BAKR6397
SDS
spec_JT6397-5
Product specification
Customers Who Bought This Also Bought