MES sodium salt ≥85.0% for biopharmaceutical production, purified, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Non-Regulated, With Information
About this item
Synonyms: Sodium 3-morpholin-1-ylethylsulphonate,MES sodium salt,Sodium 3-morpholin-1-ylethylsulfonate,2-(N-Morpholino)ethanesulphonic acid sodium salt,2-(N-Morpholino)ethanesulfonic acid sodium salt
CAS Number:71119-23-8
:MFCD31920063
Formula:C₆H₁₂NNaO₄S
MW:217.22 g/mol
MDL Number:MFCD00065473
:Length:
:Width:
:Height:
:Ø
:Thickness:
:85,0
:Management of Change (MOC) category = NI
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 217.22 g/mol
EINECS:275-203-2
:M.O.C. Category NI
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:
- Assay (C₆H₁₂NNaO₄S):≥85.0 %
- Appearance:Passes Test
- Identification (by IR):Passes Test
- pH of 1% Aqueous Solution at 25°C:8.9 - 10.1
- Solubility:Passes Test
- Ultraviolet Absorbance of a 10% Aqueous Solution - 280 nm:≤0.03
- Ultraviolet Absorbance of a 10% Aqueous Solution - 260 nm:≤0.05
- Water (H₂O):≤15.0 %
Specification Test Results
Related Information
Certificates
Search by Lot Number4813 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
sds_BAKR4813
SDS
Customers Who Bought This Also Bought