γ-(N-Morpholino)propanesulfonic acid (MOPS) ≥99% (dried basis), purified, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Certificates
About this item
Synonyms: 3-Morpholinopropane-1-sulfonic acid,3-(N-Morpholino)propanesulfonic acid,3-Morpholinopropane-1-sulphonic acid
Formula:C₇H₁₅NO₄S
MW:209.27 g/mol
Melting Pt:250 °C
MDL Number:MFCD00006183
CAS Number:1132-61-2
Merck Index:12,06346
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99
:Management of Change (MOC) category = NI
Flash Pt:110 °C (closed cup)
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: Min. 250 °C|Melting Pt: 250 °C
:Flash Pt:
:Flash Pt: 110 °C (closed cup)
:M.W. 209.27 g/mol
:γ-(N-Morpholino)propanesulphonic acid (MOPS)
Storage Temperature:Ambient
EINECS:214-478-5
:110 °C
:GHS07
:M.O.C. Category NI
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:
- Assay (C₇H₁₅NO₄S) (dried basis):≥99 %
- DNase Activity (None Detected):Passes Test
- RNase Activity:Passes Test
- Protease Activity:Passes Test
- Appearance:Passes Test
- pH of 1% Solution at 25°C:2.5 - 4.5
- Solution (10%) in Water:Passes Test
- Loss on Drying:≤1 %
- Absorbance of a 0.1M Solution (1-cm path vs water) at - 260 nm:≤0.020
- Trace Impurities - ACS - Heavy Metals (as Pb):≤5 ppm
- Trace Impurities - Iron (Fe):≤5 ppm
Specification Test Results
Related Information
Certificates
Search by Lot Number4810 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
sds_BAKR4810
SDS
Customers Who Bought This Also Bought