Magnesium sulfate heptahydrate 99.0-100.5% (by anhydrous basis), crystals USP, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Regulated
About this item
Synonyms: sulphuric acid magnesium salt heptahydrate,sulfuric acid magnesium salt heptahydrate,Epsom salt
Formula:MgSO₄·7H₂O
MW:246.48 g/mol
Melting Pt:1124 °C
Density:1.67 g/cm³
MDL Number:MFCD00149785
CAS Number:10034-99-8
Merck Index:13,05715
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,0-100,5
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 1124 °C
:Flash Pt:
:Flash Pt:
:M.W. 246.48 g/mol
Storage Temperature:Ambient
EINECS:231-298-2
:M.O.C. Category R
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Meets B.P. Chemical Specifications:
- Meets E.P. Chemical Specifications:
- GMP Manufactured Product:
- Meets U.S.P Requirements:
- Preserve in well-closed containers.:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- USP - Identification:Passes Test
- USP - pH (1 in 20):5.0 - 9.2
- USP - Loss on Ignition:40.0 - 52.0 %
- USP - Chloride (Cl):≤ 0.014 %
- USP - Iron (Fe):≤ 0.5 ppm
- USP - Selenium (Se):≤ 0.003 %
- USP - Assay (MgSO₄) (anhydrous basis):99.0 - 100.5 %
- USP - Endotoxin Concentration (EU/g):≤ 2.5
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Acidity or Alkalinity:Passes Test
- EP/BP - Chloride (Cl):≤ 300 ppm
- EP/BP - Iron (Fe):≤ 20 ppm
- EP/BP - Loss on Drying:48.0 - 52.0 %
- EP/BP - Assay (MgSO₄) (anhydrous basis):99.0 - 100.5 %
- Solution Test:Passes Test
- No Class 1,2,3 or other solvents are used or produced in the:
- manufacturing or purification of the product.:
- Elemental Impurities (USP 232, EP 5.20) - Information on:
- elemental impurities for this product is available on the:
- associated Product Regulatory Data Sheet and elemental impurity:
- profile report.:
- This product is intended for use in preparing parenteral and:
- nonparenteral dosage forms.:
Specification Test Results
Related Information
Certificates
Search by Lot Number2504 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals VWR Top Rated Guide
Product Literature
spec_JT2504-1 spec_JT2504-5 spec_JT2504-7 spec_JT2504-8
Product specification
Customers Who Bought This Also Bought