NSK-24™ Nitri-Solve® Knit Gloves, Showa
Supplier: Showa
Certificates
About this item
Chemical-resistant nitrile gloves provide protection against a wide array of solvents, oils, and acids.
- Color: blue
- They have a rough texture finish for a sure grip in wet and oily applications
The gloves offer flexibility and dexterity not available in thicker gloves and feature an interlock knit liner for an extra degree of protection and comfort.
Specifications
- Catalog No:
- 80090-036
- 80090-032
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable