Methanol ≥99.9%, BioSyn™ for biosynthesis, for applications requiring low water content, Burdick & Jackson™
Supplier: Burdick & Jackson
Certificates
About this item
Synonyms: Hydroxy methane,Carbinol,Wood alcohol
Formula:CH₃OH
MW:32.04 g/mol
Boiling Pt:64.7 °C
Melting Pt:–98 °C
Density:0.792 g/cm³ (20 °C)
Flash Pt:11 °C
MDL Number:MFCD00004595
CAS Number:67-56-1
ADR:3,II
Merck Index:13,05984
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,9
:Filtered through a 0.2 µm filter.
:Boiling Pt:
:Boiling Pt: 64.7 °C
:Melting Pt:
:Melting Pt: -98 °C|Melting Pt: –98 °C
:Flash Pt:
:Flash Pt: 11 °C
:M.W. 32.04 g/mol
EINECS:200-659-6
:-98
:64.7 °C
UN:1230
REACH:01-2119433307-44
:3
:II
:11 °C
:GHS02
:GHS08
Storage Temperature:Ambient
:Filter 0,2 um
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- GC Analysis:99.9 % min.
- Refractive Index (20ºC):1.3280 - 1.3288
- Residue:1 mg/L max.
- UV Absorbance @ 225 nm:0.16 AU max.
- UV Absorbance @ 250 nm:0.02 AU max.
- UV Absorbance @ 300 nm:0.005 AU max.
- UV Absorbance @ 400 nm:0.005 AU max.
- UV Cutoff:205 nm max.
- Water by Karl Fischer Titration:300 ppm max.
- Burdick & Jackson™, Methanol ≥99.9%, BioSyn™ for biosynthesis, for applications requiring low water content:
Specification Test Results
Related Information
Customers Who Bought This Also Bought