Urea 98.0-102.0% USP, Multi-Compendial, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Regulated
About this item
Synonyms: Carbonyl diamide,Carbamide,Diaminomethanone
Formula:NH₂CONH₂
MW:60.06 g/mol
Boiling Pt:197 °C (1013 hPa)
Melting Pt:132.7 °C
Density:1.32 g/ml (20 °C)
MDL Number:MFCD00008022
CAS Number:57-13-6
Merck Index:12,10005
:Length:
:Width:
:Height:
:Ø
:Thickness:
:98,0-102,0
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt: 197 °C (1013 hPa)
:Melting Pt:
:Melting Pt: 132.7 °C
:Flash Pt:
:Flash Pt:
:M.W. 60.06 g/mol
Storage Temperature:Ambient
EINECS:200-315-5
REACH:01-2119463277-33
:GHS07
:GHS08
:M.O.C. Category R
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Meets U.S.P Requirements:
- GMP Manufactured Product:
- Meets B.P. Chemical Specifications:
- Meets E.P. Chemical Specifications:
- Meets J.P. Chemical Specifications:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- USP - Identification A:Passes Test
- USP - Identification B:Passes Test
- USP - Assay (NH₂CONH₂):98.0 - 102.0 %
- USP - Residue on Ignition:≤0.1 %
- USP - Alcohol Insoluble Matter:≤0.04 %
- USP - Organic impurities: Urea related compound A:≤0.1 %
- USP - Organic impurities: Any individual unspecified impurity:≤0.1 %
- USP - Organic impurities: Total impurities:≤2.0 %
- USP - Endotoxin Concentration (EU/g):≤2.5
- EP/BP - Assay (NH₂CONH₂) (dried basis):98.5 - 101.5 %
- EP/BP - Identification A:Passes Test
- EP/BP - Identification B:Passes Test
- EP/BP - Appearance of Solution:Passes Test
- EP/BP - Alkalinity:Passes Test
- EP/BP - Biuret:≤0.1 %
- EP/BP - Ammonium (NH₄):≤500 ppm
- EP/BP - Loss on Drying at 105°C:≤1.0 %
- EP/BP - Ash (sulfated):≤0.1 %
- JP - Assay (NH₂CONH₂):99.0 - 101.0 %
- JP - Identification A:Passes Test
- JP - Identification B:Passes Test
- JP - Chloride (Cl):≤0.007 %
- JP - Sulfate (SO₄):≤0.010 %
- JP - Heavy Metals (as Pb):≤20 ppm
- JP - Alcohol Insoluble Matter:≤0.040 %
- JP - Residue on Ignition:≤0.10 %
- JP - Melting Point:132.5 - 134.5 °C
- No Class 1,2,3 or other solvents are used or produced in the:
- manufacturing or purification of the product.:
- Manufactured exclusively for Avantor Performance Materials, LLC., by BioSpectra, Inc., Stroudsburg, PA:
- Elemental Impurities (USP 232, EP 5.20) - Information on elemental:
- impurities for this product is available on the associated Product:
- Regulatory Data Sheet and elemental impurity profile report.:
Specification Test Results
Related Information
Certificates
Search by Lot Number4203 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
sds_BAKR4203
SDS
spec_JT4203-60 spec_JT4203-7
Product specification
Customers Who Bought This Also Bought