Dual Scale Hydrometers, Specific Gravity/Baume
Supplier: Heathrow Scientific
Certificates
About this item
Dual-Scale Hydrometers have two scales on the stem, covering different applications in one unit.
- Calibrated for use at 60 °F
The Baume scale has two different scales: one for liquids heavier than water and one for liquids lighter than water.
Customers Who Bought This Also Bought