Diisopropyl ether ≥99.0% (by GC, corrected for water content) stabilized, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Non-Regulated
About this item
Synonyms: Isopropyl ether
Formula:(CH₃)₂CHOCH(CH₃)₂
MW:102.18 g/mol
Boiling Pt:68.5 °C (1013 hPa)
Melting Pt:–86 °C
Density:0.726 g/cm³ (20 °C)
Flash Pt:–28 °C (closed cup)
MDL Number:MFCD00008880
CAS Number:108-20-3
ADR:3,II
Merck Index:13,05232
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,0
:Management of Change (MOC) category = N
:Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol) 0.01 %
:Boiling Pt:
:Boiling Pt: 68.5 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –86 °C
:Flash Pt:
:Flash Pt: –28 °C (closed cup)
:M.W. 102.18 g/mol
:Diisopropyl ether
EINECS:203-560-6
UN:1159
:3
:II
:–28 °C
:GHS02
:GHS07
Storage Temperature:Ambient
:BHT
:0.01 %
:M.O.C. Category N
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Meets ACS Reagent Chemical Requirements:
- For Laboratory,Research,or Manufacturing Use:
- Assay ((CH₃)₂CHOCH(CH₃)₂) (by GC, corrected for water):≥99.0 %
- Color (APHA):≤25
- Residue after Evaporation:≤0.0100 %
- Titrable Acid (meq/g):≤0.0007
- Peroxide (as C₆H₁₄O₂):≤0.05 %
- Stabilized with approximately 0.01% Butylated Hydroxytoluene:
- J.T.Baker®, Diisopropyl ether ≥99.0% (by GC, corrected for water content) stabilized, BAKER ANALYZED® ACS:
Specification Test Results
Related Information
Certificates
Search by Lot Number9243 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
Brand Definition Baker Analyzed
Datasheet
sds_BAKR9243
SDS
Customers Who Bought This Also Bought