UltraSense™ EC Powder-Free Nitrile Gloves, Microflex®, Ansell
Supplier: Microflex
CE Compliant
Certificates
About this item
Ultra-soft, powder-free gloves are made from a nitrile formulation that has the fit and feel of latex and provides reliable protection against many hazardous and infectious substances.
- Ambidextrous
- Beaded cuffs
- Textured fingers
Non-stick surface resists tapes and adhesives.
Caution: Components used in making these gloves may cause allergic reactions in some users. Follow your institution's policies for use.
Specifications
- Catalog No:
- CA89065-284
- CA89065-278
- CA89065-282
- CA89065-280
- CA89065-288
- CA89065-286
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Cuff thickness:
- 3.9 mil
- 3.9 mil
- 3.9 mil
- 3.9 mil
- 3.9 mil
- 3.9 mil
- Finger thickness:
- 5.9 mil
- 5.9 mil
- 5.9 mil
- 5.9 mil
- 5.9 mil
- 5.9 mil
- Palm thickness:
- 4.7 mil
- 4.7 mil
- 4.7 mil
- 4.7 mil
- 4.7 mil