PYREX® Boiling Flasks, Florence, Flat Bottom, Long Neck, with Standard Taper Joint, Corning
Supplier: Corning
Certificates
About this item
PYREX® borosilicate glass, long neck.
- High thermal shock resistance
- Provide contamination free results
- High chemical durability
These flasks have full length outer standard tapered 24/40 joint. The normal working temperature is 230 °C (446 °F).
Specifications
- Catalog No:
- 89091-254
- 89091-418
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey