MidKnight™ Nitrile Examination Gloves, Microflex®, Ansell
Supplier: Microflex
CE Compliant
Certificates
About this item
Powder-free MidKnight™ Nitrile Examination gloves provide exceptional tactile sensitivity, comfort, and durability.
- Powder-free, fully textured for firm grip
- Exceptional tactile sensitivity, comfort, and durability
- Distinct color masks stains
- Thickness: 5.5 mil (finger)
Distinct color masks stains. Fully textured for a firm grip.
Caution: Components used in making these gloves may cause allergic reactions in some users. Follow your institution's policies for use.
Specifications
- Catalog No:
- CA94001-374
- CA94001-372
- CA94001-370
- CA94001-368
- CA94001-366
- CA94001-364
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Cuff thickness:
- 3.1 mil
- 3.1 mil
- 3.1 mil
- 3.1 mil
- 3.1 mil
- 3.1 mil
- Finger thickness:
- 5.5 mil
- 5.5 mil
- 5.5 mil
- 5.5 mil
- 5.5 mil
- 5.5 mil
- Palm thickness:
- 4.7 mil
- 4.7 mil
- 4.7 mil
- 4.7 mil
- 4.7 mil
- 4.7 mil