Ammonium iron (III) citrate (14.5 - 16.0% Fe), green, powder FCC, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Regulated Limited
About this item
Synonyms: Ammonium ferric citrate,Ferric Ammonium Citrate
Formula:C₆H₅+4y)Fe(x)N(y)O₇
MDL Number:MFCD00013099
CAS Number:1185-57-5
Merck Index:13,00518
:Ammonium iron (III) citrate
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Management of Change (MOC) category = RL
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 265.00 g/mol
EINECS:214-686-6
:M.O.C. Category RL
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Meets F.C.C. Requirements:
- Food GMP Manufactured Product:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Food Chemical:
- FCC - Assay (as Fe):14.5 - 16.0 %
- FCC - Identification:Passes Test
- FCC - Ferric Citrate:Passes Test
- FCC - Lead (Pb):≤2 mg/kg
- FCC - Mercury (Hg):≤1 mg/kg
- FCC - Oxalate (C₂O₄):Passes Test
- FCC - Sulfate (SO₄):≤0.3 %
- J.T.Baker®, Ammonium iron (III) citrate (14.5 - 16.0% Fe), green, powder FCC:
Specification Test Results
Related Information
Certificates
Search by Lot Number1977 Product Specifications
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
sds_BAKR1977
SDS
Customers Who Bought This Also Bought