Esteem® TRU-BLU™ Nitrile Examination Gloves, Cardinal
Supplier: Avantor
Certificates
About this item
Stretchy, powder-free nitrile gloves feature a high coefficient of friction (grip), even when wet
- Beaded cuff resists tearing and minimizes cuff rolldown
- Microtextured for excellent grip in wet or dry applications
- Ambidextrous
- Thickness: 5.1 mil (finger)
They also provide good tactile sensitivity while maintaining film strength and barrier protection. Gloves have been tested for resistance to chemotherapy agents and for use with common hospital and laboratory chemicals.
Caution: Note: Gloves used for protection against chemotherapy drug exposure must be selected specifically for the type of drugs being used.
Specifications
- Catalog No:
- CA89040-198
- Ambidextrous/Hand-specific:
- Ambidextrous
- Disposable / reusable:
- Disposable
- Powdered/Powder-free:
- Powder-free
- Finger thickness:
- 5.1 mil