Dexi-Task Disposable Nitrile Examination Gloves, Honeywell Safety
Supplier: Honeywell Safety Products
Certificates
About this item
Ideal for food handling, packaging, inspections, and small parts handling.
- Ambidextrous gloves provide a smooth grip and more puncture resistance than rubber or vinyl gloves
- Latex-free
- Rolled cuff
Color: blue.
Certifications: Gloves comply with USDA and FDA regulations 21CFR for use in food processing.
Specifications
- Catalog No:
- CA89022-060
- CA89022-062
- CA89022-050
- CA89022-044
- CA89022-066
- CA89022-064
- CA89022-048
- CA89022-046
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Powdered/Powder-free:
- Low-powdered
- Low-powdered
- Low-powdered
- Low-powdered