PYREX® VISTA™ Micro Filtering Flasks, Corning
Supplier: Corning
Certificates
About this item
Made of borosilicate glass.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion, giving relatively high resistance to temperature changes
These small filtering flasks are recommended for microchemical use. All flasks have permanent blue enamel marking spots.
Tubulation outer Ø: 10 mm
Certifications: ASTM E-1406 Type 2
Specifications
- Catalog No:
- 89090-314
- 89090-358
- 89090-396
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey