PowerCoat® Disposable Gloves, Honeywell Safety
Supplier: Honeywell Safety Products
Certificates
About this item
Rugged 8-mil disposable nitrile gloves resist chemical splash and help protect against minor nicks, cuts, puncture, and abrasion
- Ambidextrous
- Beaded cuff
Gloves are textured for improved grip and have very good flexibility and dexterity. They are powder-free and silicone-free, making them ideal for food and high-tech applications including meat, poultry, and seafood processing, canning, food preparation, laboratory tasks, and painting applications in automotive, aerospace, furniture, and appliance manufacture.
Certifications: Gloves are manufactured in an ISO-certified facility that is also silicone-free.
Ordering information: Gloves are packaged in dispenser boxes, 50 gloves per box.
Specifications
- Catalog No:
- CA56619-951
- Ambidextrous/Hand-specific:
- Ambidextrous
- Disposable / reusable:
- Disposable
- Powdered/Powder-free:
- Powder-free