UltraSense™ Powder-Free Nitrile Gloves, Microflex®, Ansell
Supplier: Microflex
CE Compliant
Certificates
About this item
UltraSense™ powder-free nitrile gloves offer a blend of comfort and durability unique to nonlatex gloves.
- Ultra soft, comfortable fit and feel
- Reliable barrier protection against hazardous and infectious substances
- Latex free, eliminates natural rubber latex allergic reactions
- For single use only
The proprietary, ultra-soft formulation provides a comfortable fit and feel while maintaining reliable barrier protection against many hazardous and infectious substances. These highly elastic gloves conform to the hand to allow a full range of motion, thus reducing hand fatigue and stress. Textured fingers offer excellent grip in wet or dry conditions, and a polymer coating allows quick and easy donning. The nonlatex composition eliminates the possibility of natural rubber latex allergic reactions.
Caution: Components used in making these gloves may cause allergic reactions in some users. Follow your institution's policies for use.
Specifications
- Catalog No:
- CA32919-002
- CA32919-000
- CA32919-008
- CA32919-006
- CA32919-004
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Disposable / reusable:
- Disposable
- Disposable
- Disposable
- Disposable
- Disposable
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Cuff thickness:
- 2.8 mil
- 2.8 mil
- 2.8 mil
- 2.8 mil
- 2.8 mil
- Finger thickness:
- 0.13 mm/5.1 mil
- 0.13 mm/5.1 mil
- 0.13 mm/5.1 mil
- 0.13 mm/5.1 mil
- 0.13 mm/5.1 mil
- Palm thickness:
- 0.08 mm/3.2 mil
- 0.08 mm/3.2 mil
- 0.08 mm/3.2 mil
- 0.08 mm/3.2 mil
- 0.08 mm/3.2 mil