Hydrogen peroxide solution 30%, AR Select® ACS for trace metal analysis, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Non-Regulated
About this item
Formula:H₂O₂
MW:34.01 g/mol
Boiling Pt:∼107 °C (1013 hPa)
Melting Pt:< 0 °C
Density:1.11 g/cm³ (20 °C)
MDL Number:MFCD00011333
CAS Number:7722-84-1
ADR:5.1,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Management of Change (MOC) category = N
:Boiling Pt:
:Boiling Pt: ∼107 °C (1013 hPa)
:Melting Pt:
:Melting Pt: < 0 °C
:Flash Pt:
:Flash Pt:
:Density: 1.11 g/cm³ (20 °C)
:M.W. 34.01 g/mol
:Hydrogen peroxide
:34.01
Storage Temperature:Ambient
EINECS:231-765-0
:∼107 °C (1013 hPa)
UN:2014
:5.1
:II
:1.11
:GHS03
:GHS05
:GHS06
:GHS08
:M.O.C. Category N
:14,04798
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Meets ACS Reagent Chemical Requirements:
- For Laboratory,Research,or Manufacturing Use:
- ACS - Assay (H₂O₂):30.0 - 32.0 %
- ACS - Color (APHA):≤10
- ACS - Residue after Evaporation:≤20 ppm
- ACS - Free Acid (µeq/g):≤0.6
- Trace Impurities - Aluminum (Al):≤0.200 ppm
- Ammonium (NH₄):≤5 ppm
- Arsenic and Antimony (as As):≤0.010 ppm
- Trace Impurities - Barium (Ba):≤0.010 ppm
- Trace Impurities - Beryllium (Be):≤0.010 ppm
- Trace Impurities - Bismuth (Bi):≤0.010 ppm
- Trace Impurities - Boron (B):≤0.010 ppm
- Trace Impurities - Cadmium (Cd):≤0.010 ppm
- Trace Impurities - Calcium (Ca):≤0.100 ppm
- Chloride (Cl):≤2 ppm
- Trace Impurities - Chromium (Cr):≤0.040 ppm
- Trace Impurities - Cobalt (Co):≤0.010 ppm
- Trace Impurities - Copper (Cu):≤0.010 ppm
- Trace Impurities - Gallium (Ga):≤0.010 ppm
- Trace Impurities - Germanium (Ge):≤0.020 ppm
- Trace Impurities - Gold (Au):≤0.010 ppm
- ACS - Heavy Metals (as Pb):≤1 ppm
- Trace Impurities - Iron (Fe):≤0.050 ppm
- Trace Impurities - Lead (Pb):≤0.010 ppm
- Trace Impurities - Lithium (Li):≤0.010 ppm
- Trace Impurities - Magnesium (Mg):≤0.010 ppm
- Trace Impurities - Manganese (Mn):≤0.050 ppm
- Trace Impurities - Mercury (Hg):≤0.001 ppm
- Trace Impurities - Molybdenum (Mo):≤0.010 ppm
- Trace Impurities - Nickel (Ni):≤0.020 ppm
- Nitrate (NO₃):≤2 ppm
- Phosphate (PO₄):≤2 ppm
- Trace Impurities - Potassium (K):≤0.400 ppm
- Trace Impurities - Silver (Ag):≤0.050 ppm
- Trace Impurities - Sodium (Na):≤0.050 ppm
- Trace Impurities - Strontium (Sr):≤0.010 ppm
- ACS - Sulfate (SO₄):≤5 ppm
- Trace Impurities - Thallium (Tl):≤0.050 ppm
- Trace Impurities - Tin (Sn):≤0.500 ppm
- Trace Impurities - Titanium (Ti):≤0.010 ppm
- Trace Impurities - Vanadium (V):≤0.010 ppm
- Trace Impurities - Zinc (Zn):≤0.100 ppm
- Trace Impurities - Zirconium (Zr):≤0.010 ppm
- Macron Fine Chemicals™, Hydrogen peroxide solution 30%, AR Select® ACS for trace metal analysis:
Specification Test Results
Related Information
Certificates
Search by Lot NumberAvantor Portfolio of Chemicals
Product Literature
sds_MACRV340
SDS
V340 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Customers Who Bought This Also Bought