Hexane (mixture of isomers) ≥98.5% (by GC), AR® ACS, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Certificates
About this item
Synonyms: Hexanes
CAS Number:110-54-3
:92112-69-1
Formula:H₃C(CH₂)₄CH₃
MW:86.18 g/mol
Boiling Pt:68 - 70 °C (1013 hPa)
Melting Pt:-95 °C
Density:0.66 g/cm³ (20 °C)
Flash Pt:–23 °C
MDL Number:MFCD02179311
ADR:3,II
Merck Index:13,04712
:Length:
:Width:
:Height:
:Ø
:Thickness:
:98,5
:Management of Change (MOC) category = N
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: -95 °C
:Flash Pt:
:Flash Pt: –23 °C
:M.W. 86.18 g/mol
EINECS:203-777-6
:-95
UN:1208
:3
:II
:–23 °C
:GHS02
:GHS08
:GHS07
:GHS09
Storage Temperature:Ambient
:M.O.C. Category N
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Meets ACS Reagent Chemical Requirements:
- For Laboratory,Research,or Manufacturing Use:
- ACS - Assay (sum of 5 isomers, total hexanes, plus methylcyclopentane)(by GC):≥98.5 %
- ACS - Assay (as n-hexane) (by GC), For Information Only:
- Appearance (clear, colorless liquid):Passes Test
- ACS - Color (APHA):≤10
- ACS - Residue after Evaporation:≤0.001 %
- ACS - Sulfur Compounds (as S):≤0.005 %
- ACS - Thiophene:Passes Test
- ACS - Water-Soluble Titrable Acid (meq/g):≤0.0003
- Macron Fine Chemicals™, Hexane (mixture of isomers) ≥98.5% (by GC), AR® ACS:
Specification Test Results
Related Information
Certificates
Search by Lot Number5189 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Avantor Portfolio of Chemicals
Product Literature
Macron 5189.pdf
Datasheet
sds_MACR5189
SDS
Customers Who Bought This Also Bought