Hydrochloric acid 36.5% - 38.0% NF, FCC, ACS, Macron Fine Chemicals™
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Regulated
About this item
Synonyms: Muriatic acid
Formula:HCl
MW:36.46 g/mol
Boiling Pt:110 °C (1013 hPa)
Melting Pt:–30 °C
Density:1.18 g/cm³ (20 °C)
MDL Number:MFCD00011324
CAS Number:7647-01-0
ADR:8,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt: 110 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –30 °C
:Flash Pt:
:Flash Pt:
:M.W. 36.46 g/mol
:36.46
EINECS:231-595-7
UN:1789
:8
:II
:GHS05
:GHS07
Storage Temperature:Ambient
:M.O.C. Category R
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Food GMP Manufactured Product:
- Meets F.C.C. Requirements:
- GMP Manufactured Product:
- Meets N.F. Requirements:
- Meets ACS Reagent Chemical Requirements:
- Store below 25°C:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Food Chemical:
- Bulk Pharmaceutical Chemical:
- NF - Assay (HCl)(NF):36.5 - 38.0 %
- NF - Bromide or Iodide:Passes Test
- NF - Free Bromine or Chlorine:Passes Test
- NF - Identification:Passes Test
- NF - Residue on Ignition:≤ 0.008 %
- NF - Sulfate (SO₄):Passes Test
- NF - Sulfite (SO₃):Passes Test
- FCC - Assay (HCl)(FCC):36.5 - 38.0 %
- FCC - Color (APHA):Passes Test
- FCC - Baume Degrees:22.7 - 23.4
- FCC - Identification:Passes Test
- FCC - Iron (Fe) (mg/kg):≤ 5
- FCC - Lead (Pb) (mg/kg):≤ 1
- FCC - Mercury (Hg)(mg/kg):≤ 0.10
- FCC - Nonvolatile Residue:≤ 0.5 %
- Total Organic Compounds - Non-Fluorine Containing (mg/kg):≤ 5
- Total Organic Compounds - Benzene (mg/kg):≤ 0.05
- Total Organic Compounds - Fluorinated Organic Compounds(mg/kg):≤ 25
- Total Organic Compounds - Oxidizing Substances:≤ 0.003 %
- Reducing Substances:≤ 0.007 %
- Specific Gravity:1.1856 - 1.1924
- Sulfate (SO₄):≤ 0.5 %
- ACS - Assay (HCl):36.5 - 38.0 %
- ACS - Ammonium (NH₄):≤ 3 ppm
- ACS - Arsenic (As):≤ 0.010 ppm
- ACS - Bromide (Br):≤ 0.005 %
- ACS - Color (APHA):≤ 10
- ACS - Extractable Organic Substances:≤ 5 ppm
- ACS - Free Chlorine (as Cl):≤ 1 ppm
- ACS - Heavy Metals (as Pb):≤ 1 ppm
- Trace Impurities - ACS - Iron (Fe):≤ 0.200 ppm
- ACS - Residue after Ignition:≤ 5 ppm
- ACS - Sulfate (SO₄):≤ 1 ppm
- ACS - Sulfite (SO₃):≤ 1 ppm
- Appearance:Passes Test
- No Class 1,2,3 or other solvents are used or produced in the:
- manufacturing or purification of the product.:
- Elemental Impurities (USP 232, EP 5.20) - Information on elemental:
- impurities for this product is available on the associated Product:
- Regulatory Data Sheet and elemental impurity profile report.:
- Macron Fine Chemicals™, Hydrochloric acid 36.5% - 38.0% NF, FCC, ACS:
Specification Test Results
Related Information
Certificates
Search by Lot Number2062 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Macron 2062.pdf
Datasheet
sds_MACR2062
SDS
spec_MK206204 spec_MK206246
Product specification
Customers Who Bought This Also Bought