n-Heptane ≥99.0% (by GC, corrected for water content), BAKER ANALYZED®, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Non-Regulated
About this item
Formula:H₃C(CH₂)₅CH₃
MW:100.20 g/mol
Boiling Pt:98.4 °C (1013 hPa)
Melting Pt:–90.6 °C
Density:0.6838 g/cm³ (20 °C)
Flash Pt:–4 °C
MDL Number:MFCD00009544
CAS Number:142-82-5
ADR:3,II
Merck Index:13,04679
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,0
:Management of Change (MOC) category = N
:Boiling Pt:
:Boiling Pt: 98.4 °C (1013 hPa)
:Melting Pt:
:Melting Pt: -90.6 °C|Melting Pt: –90.6 °C
:Flash Pt:
:Flash Pt: –4 °C
:M.W. 100.20 g/mol
Storage Temperature:Ambient
EINECS:205-563-8
UN:1206
:3
:II
:–4 °C
:GHS02
:GHS08
:GHS07
:GHS09
:M.O.C. Category N
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:
- Assay (CH₃(CH₂)₅CH₃) (by GC, corrected for water):≥99.0 %
- Color (APHA):≤10
- Residue after Evaporation:≤0.001 %
- J.T.Baker®, n-Heptane ≥99.0% (by GC, corrected for water content), BAKER ANALYZED®:
Specification Test Results
Related Information
Certificates
Search by Lot NumberAvantor Portfolio of Chemicals VWR Top Rated Guide VWR® Chemicals in Poly-Coated Glass Bottles
Product Literature
Brand Definition Baker Analyzed
Datasheet
M956 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Customers Who Bought This Also Bought