Hydrofluoric acid 48.0 - 51.0% (by acidimetry), BAKER ANALYZED® ACS, J.T.Baker®
Supplier: Avantor
MOC: Non-Regulated
About this item
Formula:HF
MW:20.01 g/mol
MDL Number:MFCD00011346
CAS Number:7664-39-3
:20.01
ADR:8,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Management of Change (MOC) category = N
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 20.01 g/mol
Storage Temperature:Ambient
EINECS:231-634-8
UN:1790
:8
:II
:GHS06
:GHS05
:GHS08
:M.O.C. Category N
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Meets ACS Reagent Chemical Requirements:
- For Laboratory,Research,or Manufacturing Use:
- Assay (HF) (by acidimetry):48.0 - 51.0 %
- Fluosilicic Acid (H₂SiF₆):≤0.01 %
- Residue after Ignition:≤5 ppm
- Chloride (Cl):≤5 ppm
- Nitrate (NO₃):≤5 ppm
- Phosphate (PO₄):≤1 ppm
- Sulfate and Sulfite (as SO₄):≤5 ppm
- Trace Impurities - Arsenic and Antimony (as As):≤50.0 ppb
- Trace Impurities - Copper (Cu):≤50 ppb
- Heavy Metals (as Pb):≤500.0 ppb
- Trace Impurities - Iron (Fe):≤500.0 ppb
- Trace Impurities - Lead (Pb):≤100 ppb
- Trace Impurities - Nickel (Ni):≤50 ppb
- Meets Reagent Specifications for testing USP/NF monographs:
- J.T.Baker®, Hydrofluoric acid 48.0 - 51.0% (by acidimetry), BAKER ANALYZED® ACS:
Specification Test Results
Related Information
Certificates
Search by Lot Number9560 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Customers Who Bought This Also Bought