Hexane (mixture of isomers) ≥99.5% (by GC, corrected for water content) (n-hexane ≥95%), ULTRA RESI-ANALYZED® for organic residue analysis, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Non-Regulated, With Information
About this item
Synonyms: Hexanes
CAS Number:110-54-3
:92112-69-1
Formula:H₃C(CH₂)₄CH₃
MW:86.18 g/mol
Boiling Pt:69 °C (1013 hPa)
Melting Pt:–94.3 °C
Density:0.659 g/cm³ (20 °C)
Flash Pt:–23 °C
MDL Number:MFCD02179311
ADR:3,II
Merck Index:13,04712
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,5
:Management of Change (MOC) category = NI
:Boiling Pt:
:Boiling Pt: 69 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –94.3 °C
:Flash Pt:
:Flash Pt: –23 °C
:M.W. 86.18 g/mol
Storage Temperature:Ambient
EINECS:203-777-6
UN:1208
:3
:II
:–23 °C
:GHS02
:GHS08
:GHS07
:GHS09
:M.O.C. Category NI
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:
- FID-Sensitive Impurities (as 2-Octanol) Single Impurity Peak (ng/mL):≤5
- ECD Sensitive Impurities (as Heptachlor Epoxide) Single Peak (pg/mL):≤10
- ECD-Sensitive Impurities (as Ethylene Dibromide) - Single Impurity Peak (ng/mL):≤5
- Assay (Total Saturated C₆ Isomers) (by GC, corrected for water):≥99.5 %
- Assay (as n-Hexane) (by GC, corrected for water):≥95 %
- Color (APHA):≤10
- Residue after Evaporation:≤1.0 ppm
- Substances Darkened by H₂SO₄:Passes Test
- Water (by KF, coulometric):≤0.05 %
- MEETS SPECIFICATIONS WITHIN THE EXPIRATION PERIOD:
- J.T.Baker®, Hexane (mixture of isomers) ≥99.5% (by GC, corrected for water content) (n-hexane ≥95%), ULTRA RESI-ANALYZED® for organic residue analysis:
Specification Test Results
Related Information
Certificates
Search by Lot Number9262 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Avantor High-Purity Solvents and Reagents Avantor Portfolio of Chemicals VWR Top Rated Guide
Product Literature
spec_JT9262-2 spec_JT9262-3 spec_JT9262-S5
Product specification
Customers Who Bought This Also Bought