Hydrochloric acid 32 - 35%, ULTREX® II, Ultrapure for trace metal analysis, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Non-Regulated
About this item
Synonyms: Muriatic acid
Formula:HCl
MW:36.46 g/mol
Boiling Pt:110 °C (1013 hPa)
Melting Pt:–30 °C
Density:1.18 g/cm³ (20 °C)
MDL Number:MFCD00011324
CAS Number:7647-01-0
ADR:8,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Management of Change (MOC) category = N
:Boiling Pt:
:Boiling Pt: 110 °C (1013 hPa)
:Melting Pt:
:Melting Pt: –30 °C
:Flash Pt:
:Flash Pt:
:M.W. 36.46 g/mol
:36.46
EINECS:231-595-7
UN:1789
:8
:II
:GHS05
:GHS07
Storage Temperature:Ambient
:M.O.C. Category N
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:
- Assay (HCl)(w/w):32 - 35 %
- Trace Impurities - Aluminum (Al):≤20 ppt
- Trace Impurities - Antimony (Sb):≤20 ppt
- Trace Impurities - Arsenic (As):≤50 ppt
- Trace Impurities - Barium (Ba):≤10 ppt
- Trace Impurities - Beryllium (Be):≤10 ppt
- Trace Impurities - Bismuth (Bi):≤10 ppt
- Trace Impurities - Boron (B):≤100 ppt
- Trace Impurities - Cadmium (Cd):≤10 ppt
- Trace Impurities - Calcium (Ca):≤10 ppt
- Trace Impurities - Cerium (Ce):≤10 ppt
- Trace Impurities - Cesium (Cs):≤10 ppt
- Trace Impurities - Chromium (Cr):≤10 ppt
- Trace Impurities - Cobalt (Co):≤10 ppt
- Trace Impurities - Copper (Cu):≤10 ppt
- Trace Impurities - Dysprosium (Dy):≤1 ppt
- Trace Impurities - Erbium (Er):≤1 ppt
- Trace Impurities - Europium (Eu):≤1 ppt
- Trace Impurities - Gadolinium (Gd):≤1 ppt
- Trace Impurities - Gallium (Ga):≤10 ppt
- Trace Impurities - Gold (Au):≤50 ppt
- Trace Impurities - Hafnium (Hf):≤10 ppt
- Trace Impurities - Holmium (Ho):≤1 ppt
- Trace Impurities - Indium (In):≤1 ppt
- Trace Impurities - Iron (Fe):≤10 ppt
- Trace Impurities - Lanthanum (La):≤1 ppt
- Trace Impurities - Lead (Pb):≤10 ppt
- Trace Impurities - Lithium (Li):≤10 ppt
- Trace Impurities - Lutetium (Lu):≤10 ppt
- Trace Impurities - Magnesium (Mg):≤10 ppt
- Trace Impurities - Manganese (Mn):≤10 ppt
- Trace Impurities - Mercury (Hg):≤50 ppt
- Trace Impurities - Molybdenum (Mo):≤10 ppt
- Trace Impurities - Neodymium (Nd):≤1 ppt
- Trace Impurities - Nickel (Ni):≤20 ppt
- Trace Impurities - Niobium (Nb):≤1 ppt
- Trace Impurities - Palladium (Pd), For Information Only:
- Trace Impurities - Platinum (Pt), For Information Only:
- Trace Impurities - Potassium (K):≤10 ppt
- Trace Impurities - Praseodymium (Pr):≤1 ppt
- Trace Impurities - Rhenium (Re):≤10 ppt
- Trace Impurities - Rhodium (Rh):≤10 ppt
- Trace Impurities - Rubidium (Rb):≤10 ppt
- Trace Impurities - Ruthenium (Ru):≤10 ppt
- Trace Impurities - Samarium (Sm):≤1 ppt
- Trace Impurities - Scandium (Sc):≤10 ppt
- Trace Impurities - Selenium (Se), For Information Only:
- Trace Impurities - Silver (Ag):≤10 ppt
- Trace Impurities - Sodium (Na):≤10 ppt
- Trace Impurities - Strontium (Sr):≤10 ppt
- Trace Impurities - Tantalum (Ta), For Information Only:
- Trace Impurities - Tellurium (Te):≤1 ppt
- Trace Impurities - Terbium (Tb):≤1 ppt
- Trace Impurities - Thallium (Tl):≤10 ppt
- Trace Impurities - Thorium (Th):≤1 ppt
- Trace Impurities - Thulium (Tm):≤1 ppt
- Trace Impurities - Tin (Sn):≤20 ppt
- Trace Impurities - Titanium (Ti):≤10 ppt
- Trace Impurities - Tungsten (W):≤10 ppt
- Trace Impurities - Uranium (U):≤1 ppt
- Trace Impurities - Vanadium (V):≤10 ppt
- Trace Impurities - Ytterbium (Yb):≤1 ppt
- Trace Impurities - Yttrium (Y):≤1 ppt
- Trace Impurities - Zinc (Zn):≤10 ppt
- Trace Impurities - Zirconium (Zr):≤10 ppt
- Certificate Provided Reports Actual Lot Analysis:
- J.T.Baker®, Hydrochloric acid 32 - 35%, ULTREX® II, Ultrapure for trace metal analysis:
Specification Test Results
Related Information
Certificates
Search by Lot Number6900 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Avantor High-Purity Acids Avantor High-Purity Solvents and Reagents Avantor Portfolio of Chemicals Avantor™ High-Purity Acids Production Insight Solutions for Controlled Environments and Manufacturing Processes
Product Literature
More
JTBaker_6900_Datasheet.pdf
Datasheet
sds_BAKR6900
SDS
Customers Who Bought This Also Bought