Hydrogen peroxide 30% stabilized, Electronic Grade, J.T.Baker®
Catalog #: CAJT2204-1
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
CAS Number: 7722-84-1
MOC: RegulatedCertificates
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:HPPE poly bottle, pressure bottom
- Size:500 ml
Specifications
Specification Test Results
- For Microelectronic Use:
- Assay (H₂O₂):30.0 - 32.0 %
- Color (APHA):≤10
- Free Acid (µeq/g):≤0.2
- Residue after Evaporation:≤15 ppm
- Ammonium (NH₄):≤3 ppm
- Chloride (Cl):≤2 ppm
- Nitrate (NO₃):≤2 ppm
- Phosphate (PO₄):≤2 ppm
- ACS - Sulfate (SO₄):≤5 ppm
- Trace Impurities - Aluminum (Al):≤200.0 ppb
- Trace Impurities - Antimony (Sb):≤10.0 ppb
- Trace Impurities - Arsenic (As):≤10.0 ppb
- Arsenic and Antimony (as As):≤10.0 ppb
- Trace Impurities - Barium (Ba):≤20.0 ppb
- Trace Impurities - Beryllium (Be):≤10.0 ppb
- Trace Impurities - Bismuth (Bi):≤20.0 ppb
- Trace Impurities - Boron (B):≤10.0 ppb
- Trace Impurities - Cadmium (Cd):≤10.0 ppb
- Trace Impurities - Calcium (Ca):≤50.0 ppb
- Trace Impurities - Chromium (Cr):≤20.0 ppb
- Trace Impurities - Cobalt (Co):≤10.0 ppb
- Trace Impurities - Copper (Cu):≤10.0 ppb
- Trace Impurities - Gallium (Ga):≤20.0 ppb
- Trace Impurities - Germanium (Ge):≤10.0 ppb
- Trace Impurities - Gold (Au):≤10.0 ppb
- Heavy Metals (as Pb):≤500.0 ppb
- Trace Impurities - Iron (Fe):≤50.0 ppb
- Trace Impurities - Lead (Pb):≤10.0 ppb
- Trace Impurities - Lithium (Li):≤10.0 ppb
- Trace Impurities - Magnesium (Mg):≤10.0 ppb
- Trace Impurities - Manganese (Mn):≤10.0 ppb
- Trace Impurities - Molybdenum (Mo):≤10.0 ppb
- Trace Impurities - Nickel (Ni):≤10.0 ppb
- Trace Impurities - Niobium (Nb):≤10.0 ppb
- Trace Impurities - Potassium (K):≤600.0 ppb
- Trace Impurities - Silicon (Si):≤100.0 ppb
- Trace Impurities - Silver (Ag):≤10.0 ppb
- Trace Impurities - Sodium (Na):≤500.0 ppb
- Trace Impurities - Strontium (Sr):≤10.0 ppb
- Trace Impurities - Tantalum (Ta):≤10.0 ppb
- Trace Impurities - Thallium (Tl):≤50.0 ppb
- Trace Impurities - Tin (Sn):≤1000.0 ppb
- Trace Impurities - Titanium (Ti):≤10.0 ppb
- Trace Impurities - Vanadium (V):≤10.0 ppb
- Trace Impurities - Zinc (Zn):≤50 ppb
- Trace Impurities - Zirconium (Zr):≤10.0 ppb
Specification Test Results
About this item
Boiling Pt:∼107 °C (1013 hPa)
Melting Pt:<0 °C
Density:1.11 g/cm³ (20 °C)
ADR:5.1,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt: ∼107 °C (1013 hPa)
:Melting Pt:
:Melting Pt: -0.41 °C|Melting Pt: <0 °C
:Flash Pt:
:Flash Pt:
:Density: 1.11 g/cm³ (20 °C)
:M.W. 34.01 g/mol
:Hydrogen peroxide
:34.01
EINECS:231-765-0
:∼107 °C (1013 hPa)
UN:2014
:5.1
:II
:1.11
:GHS03
:GHS05
:GHS06
:GHS08
Storage Temperature:Ambient
:stab
:M.O.C. Category R
:14,04798
Formula:H₂O₂
MW:34.01 g/mol
MDL Number:MFCD00011333
CAS Number:7722-84-1
Customers Who Bought This Also Bought