Hydrogen peroxide 30% stabilized, BAKER ANALYZED® ACS, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Certificates
About this item
Formula:H₂O₂
MW:34.01 g/mol
Boiling Pt:107 °C (1013 hPa)
Melting Pt:? 0 °C
Density:1.11 g/cm³ (20 °C)
MDL Number:MFCD00011333
CAS Number:7722-84-1
ADR:5.1 (8),II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Management of Change (MOC) category = N
:Boiling Pt:
:Boiling Pt: 107 °C (1013 hPa)
:Melting Pt:
:Melting Pt: -0.41 °C|Melting Pt: ? 0 °C
:Flash Pt:
:Flash Pt:
:Density: 1.11 g/cm³ (20 °C)
:M.W. 34.01 g/mol
:Hydrogen peroxide
:34.01
EINECS:231-765-0
:-0.41
:107 °C (1013 hPa)
UN:2014
:5.1 (8)
:II
:1.11
:GHS03
:GHS05
:GHS06
:GHS08
Storage Temperature:Ambient
:stab
:M.O.C. Category N
:14,04798
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- Meets ACS Reagent Chemical Requirements:
- For Laboratory,Research,or Manufacturing Use:
- Assay (H₂O₂):30.0 - 32.0 %
- Color (APHA):≤10
- Residue after Evaporation:≤0.0020 %
- Titrable Acid (meq/g):≤0.0006
- Chloride (Cl):≤2 ppm
- Nitrate (NO₃):≤2 ppm
- Phosphate (PO₄):≤2 ppm
- ACS - Sulfate (SO₄):≤5 ppm
- Ammonium (NH₄):≤5 ppm
- Trace Impurities - Copper (Cu):≤50 ppb
- Trace Impurities - Iron (Fe):≤100.0 ppb
- Heavy Metals (as Pb):≤1000.0 ppb
- Trace Impurities - Lead (Pb):≤100 ppb
- Trace Impurities - Nickel (Ni):≤50 ppb
- Meets Reagent Specifications for testing USP/NF monographs:
- J.T.Baker®, Hydrogen peroxide 30% stabilized, BAKER ANALYZED® ACS:
Specification Test Results
Related Information
Certificates
Search by Lot Number2186 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
JTBaker_2186_Datasheet.pdf
Datasheet
sds_BAKR2186
SDS
Customers Who Bought This Also Bought