Hydrogen peroxide 3% in aqueous solution stabilized, BAKER ANALYZED®, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Non-Regulated, With Information
About this item
Formula:H₂O₂
MW:34.01 g/mol
Boiling Pt:100 °C (1013 hPa)
Density:1.01 g/cm³ (20 °C)
MDL Number:MFCD00011333
CAS Number:7722-84-1
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Management of Change (MOC) category = NI
:Boiling Pt:
:Boiling Pt: 100 °C (1013 hPa)
:Melting Pt:
:Melting Pt: -0.41 °C
:Flash Pt:
:Flash Pt:
:Density: 1.11 g/cm³ (20 °C)
:M.W. 34.01 g/mol
:Hydrogen peroxide
:34.01
Storage Temperature:Ambient
EINECS:231-765-0
:-0.41
:1.11
:M.O.C. Category NI
:14,04798
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- For Laboratory,Research,or Manufacturing Use:
- Assay (H₂O₂):3.0 - 3.5 %
- Residue after Evaporation:≤0.0500 %
- Free Acid (as H₂SO₄):≤0.01 %
- Phosphate (PO₄):≤0.003 %
- Sulfate (SO₄):≤0.005 %
- Preservative (acetophenetidin):≤0.05 %
- Chloride (Cl):≤5 ppm
- Trace Impurities - Arsenic (As):≤0.100 ppm
- ACS - Heavy Metals (as Pb):≤1 ppm
- Trace Impurities - Iron (Fe):≤5 ppm
- in aqueous solution:
Specification Test Results
Related Information
Certificates
Search by Lot Number2180 Product Specifications
Additional Product Details
Brand Definition Baker Analyzed
Datasheet
Customers Who Bought This Also Bought