Hydrochloric acid 2.0 N, Biotech Reagent, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
MOC: Regulated
About this item
Synonyms: Muriatic acid
Formula:HCl
MW:36.46 g/mol
Density:1.03 g/cm³
MDL Number:MFCD00011324
CAS Number:7647-01-0
ADR:8,II
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Management of Change (MOC) category = R
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Flash Pt:
:Flash Pt:
:M.W. 36.46 g/mol
:36.46
Storage Temperature:Ambient
EINECS:231-595-7
UN:1789
:8
:II
:GHS05
:GHS07
:M.O.C. Category R
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- GMP Manufactured Product:
- CAUTION: For Manufacturing, processing or repackaging:
- Appearance (Clear, colorless liquid):Passes Test
- Normality:1.98 - 2.02
- Ammonium (NH₄):≤ 3 ppm
- Trace Impurities - Heavy Metals:≤ 0.5 ppm
- Trace Impurities - Iron (Fe):≤ 0.1 ppm
- Sulfate (SO₄):≤ 1 ppm
- Made from U.S.P. Purified Water and Hydrochloric Acid, N.F. (Multi-Compendial) which meets BP, Ph.Eur. and JP Chemical Specifications:
- No Class 1,2,3 or other solvents are used or produced in the manufacturing or purification of the product.:
- Filtered through a 0.2 micron filter.:
- J.T.Baker®, Hydrochloric acid 2.0 N, Biotech Reagent:
Specification Test Results
Related Information
Certificates
Search by Lot Number0336 Product Specifications VWR GHS Compliance Letter (US)
Additional Product Details
Avantor Portfolio of Chemicals VWR Top Rated Guide
Product Literature
Brand Definition Baker Analyzed
Datasheet
sds_BAKR0336
SDS
spec_JT0336-3
Product specification
Customers Who Bought This Also Bought