Cell Culture Storage Bottles, Polystyrene, Corning
Supplier: Corning
Certificates
About this item
Disposable polystyrene bottles for storage of media, buffers and other aqueous solutions.
- Low profile, easy grip style has sides that facilitate handling
- Plug seal caps (45 mm) provide an airtight seal and help minimise the risk of contamination
- Bottles can be used with CorningĀ® vacuum filter systems
- Sterile, certified non-pyrogenic
Specifications
- Catalog No:
- 29442-870
- 29442-878
- 29442-874
- 28199-760
- 29442-882
- 28199-758
- 29443-026
- 28199-762
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta
- RECs, ISO 14001 - Oneonta
Specifications
Related Information
Certificates
Search by Lot Number28199-758_14819008 28199-758_36119020 28199-760_00620001 28199-760_10219003 28199-762_34719022
Conformity declaration
More
Corning Plastic-Glass-Storage-Bottles Product-Selection-Guide Corning_Plastic_Storage_Bottles_Product_Selection_Guide.pdf CorningĀ® Bottle Top Dispensers CorningĀ® Shopping list for returning to the bench
Product Literature
Corning Special Offer
Promotion Material
Customers Who Bought This Also Bought