PYREX® Distilling Flask, Vertical Necks, Outer Standard Taper Joints, 4 Necks, Corning
Supplier: Corning
Certificates
About this item
PYREX® borosilicate glass, with vertical side necks, clear.
- Very good chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
Three vertical side necks placed 90° apart at the same height with 24/40 standard taper outer joints and a larger center neck with a vertical 34/45 standard taper joint. Matt white labelling area.
Specifications
- Catalog No:
- 29215-030
- 29215-024
- 29215-026
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey