PYREX® Distilling Flasks, Angle Type, 3 Necks, [ST] Joints, Corning
Supplier: Corning
Certificates
About this item
PYREX® borosilicate glass, with angled side necks, clear.
- Excellent chemical resistance
- High temperature resistance
- Minimal thermal expansion giving relatively high resistance to temperature changes
One center neck and two side necks. Three necks with two angled side necks at the same height with 24/40 standard taper outer joints and a larger center neck with a vertical 34/45 standard taper joint. Matt white labelling area.
Specifications
- Catalog No:
- 89091-080
- 32645-392
- 89091-430
- 32645-390
- 29281-131
- 32645-391
- 89091-216
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey
- I-RECs, ISO 14001 - Monterrey