Removable Autoclave-Resistant Thermal Transfer Labels
Supplier: GA INTERNATIONAL INC
Certificates
About this item
These labels are intended for identifying metal, plastic, and glass surfaces, including surgical instruments, glassware, trays, blister packs, and other items that will undergo sterilization.
- Temperature range: −60 to 150 °C (−76 to 302 °F)
- Glove-friendly adhesive won’t stick to gloves
- Labels are readily removable without leaving behind any adhesive residue
- Can resist elevated temperatures
- Ideal for autoclave sterilization cycles at ideal for multiple autoclave sterilization cycles, gamma irradiation (up to 50 KGy), and Ethylene oxide sterilization
- Printable with all major thermal-transfer printer brands
Removable autoclave-resistant labels are intended for use in steam autoclaves, ovens, gamma irradiators, and ethylene oxide sterilization. These durable autoclave thermal transfer labels will not leave any adhesive residue after removal. They are weather and water-proof, alcohol-resistant, and have been tested to endure up to 20 autoclave cycles (at 121 °C/250 °F, 16.5 psi for 20 minutes). Compatible with all major brands of thermal transfer printers.
Customers Who Bought This Also Bought